Learn More
Innovative Research Inc Pooled Human Complement Serum 25ml
Supplier:  Innovative Research Inc ICSER25ML
Encompass_Preferred
Human Complement Serum from Innovative Research is a complex mix of serum proteins which are activated sequentially when antibodies in blood with immunity interact with the corresponding antigens. Complement proteins will quickly degrade if the serum is not prepared and stored correctly. Our complement-preserved serum is freshly collected aseptically and kept cold, and then flash-frozen at -80°C to prevent degradation of complement activity. We do not recommend this product be thawed for aliquoting, but we are able to accommodate custom volumes and aliquots upon request. 
Please note that this product does not allow for donor requests off-the-shelf, but we can create custom pooled lots to your exact specifications upon request. Contact us for details & pricing.  
Viral Testing
Each unit is tested for the standard FDA-requ
Catalog No. 50-203-6407
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.