Learn More
XenoTech,LLC Pooled Human Intestinal Microsomes, 10 mg/mL Protein Concentration, PMSF Free, 150 µL
SDP
Supplier:  XenoTech,LLC H0610INP
Encompass_Preferred
Pooled Human Intestinal Microsomes from SEKISUI XENOTECH LLC (now BioIVT) facilitate in vitro DMPK studies, providing a vital model for intestinal first-pass metabolism. They offer detailed insights for metabolite identification and enzyme activity profiling, crucial for drug-drug interactions research. The product is PMSF-free, stable up to 48 hours post-thaw at 4°C, with a protein concentration of 10 mg/mL, making it an optimal choice for various pharmacokinetic applications.
Catalog No. NC1502471