Learn More
Ace Glass, Inc. POR C ADJ FILT FUNNEL C POR C ADJ FILT FUNNEL C
SDP
Supplier:  Ace Glass, Inc. 972720
Encompass_Preferred
Microscale, vacuum adapter with vertically adjustable Hirsch filter funnel that is suitable for use with all Microscale flasks and vials with standard taper 14/10 joints. This two-piece vacuum adapter features a #7 Ace-Thred at the top with a nylon bushing and O-Ring that forms a compression seal with funnel stem, bottom joint, and side hose connection. The plain stem funnel has a disc diameter of 10 mm with a capacity above disc of approximately 7 mL. Sintered disc is Porosity C (25-50 micron). Note: without funnel, vacuum adapter will accept thermometers and bleed tubes. Complete item consists of vacuum adapter with nylon bushing and FETFE O-Ring, and funnel. Use with 5/16" I.D. tubing, size A hose connection.
Catalog No. 50-977-109