Learn More
Cole-Parmer Cole-Parmer Buchner Funnel, porcelain, 600 mL, 1/ea
Supplier:  Cole-Parmer 1780011
Encompass_Preferred
Funnel includes fixed perforated plate and is glazed inside and out, except for the rim. Resistance to acids and alkalies is excellent, except for hydrofluoric acid. Autoclavable.
Catalog No. NC1745949