Learn More
Raybiotech Inc Porcine (pig) IFN-gamma ELISA Kit
Supplier:  Raybiotech Inc ELPIFNG1
Encompass_Preferred
For cell culture supernatants, EDTA and/or Citrate treated plasma, and serum samples. Heparin is not recommended as an anticoagulant For use in this assay. Sensitivity: 500 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The sandwich ELISA antibody pair detects Porcine IFN-gamma.
Catalog No. 50-195-0187