Learn More
Bioworld PORCINE INTERLEUKIN-1 ALPHA
Supplier:  Bioworld 220606301
Encompass_Preferred
PORCINE INTERLEUKIN-1 ALPHA (RECOMBINANT): Recombinant Porcine Interleukin-1 alpha Source: E.Coli Background : A soluble factor produced by monocytes, macrophages, and other cells which activates T-lymphocytes and potentiates their r...
Catalog No. 50-253-729