Learn More
American Research Products Inc Porcine uPA ELISA Kit
Supplier:  American Research Products Inc RDRUPAP
Encompass
This Porcine Plasminogen Activator Urokinase (uPA) ELISA Kit is a precoated Sandwich ELISA assay designed to detect and quantify the target analyte in serum plasma and other biological fluids This product is for Research Use Only
Catalog No. 50-004-56697