Learn More
Medchemexpress LLC Porcn-IN-1 | 2036044-77-4 | 99.57% | 410.44 | 1 ML

Supplier: Medchemexpress LLC HY11147210MM/1ML
Porcn-IN-1 is a potent porcupine inhibitor with an IC50 of 0.5±0.2 nM. It potently inhibits the secretion of Wnt3A, confirming its role as a porcupine inhibitor. Porcupine is an enzyme that catalyzes the addition of palmitoleate to a serine residue in Wnt proteins, a process essential for Wnt protein secretion. Porcn-IN-1 is as potent as the clinical compound LGK974 in a cell-based STF reporter gene assay.
- Potent porcupine inhibitor: IC50 of 0.5±0.2 nM.
- Confirmed inhibition: Potently inhibits the secretion of Wnt3A.
- Comparable to clinical compound: As potent as LGK974 in a cell-based STF reporter gene assay.
- Moderate clearance: Demonstrates moderate clearance in human and rat liver microsomes (57 mL/min/kg and 24 mL/min/kg respectively).
- High clearance in mouse microsomes: Exhibits high clearance in mouse microsomes (109 mL/min/kg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.