Learn More
Medchemexpress LLC Porcn-IN-1 | 2036044-77-4 | 99.6% | 410.44 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11147250MG
Encompass_Preferred
Porcn-IN-1 is a potent porcupine inhibitor with an IC50 of 0.5±0.2 nM. It potently inhibits the secretion of Wnt3A. Porcupine is an enzyme that catalyzes the addition of palmitoleate to a serine residue in Wnt proteins, a process required for their secretion.
  • Potent porcupine inhibitor
  • Inhibits secretion of Wnt3A
  • Targets Porcupine and is involved in the Stem Cell/Wnt pathway
  • As potent as clinical compound LGK974 in cell-based STF reporter gene assay
  • Demonstrates moderate clearance in human and rat liver microsomes
  • Exhibits high clearance with mouse microsomes
Catalog No. 50-003-45699