Learn More
Medchemexpress LLC Porcn-IN-1 | 2036044-77-4 | MFCD31630804 | 99.6% | 410.44 | C25H19FN4O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1114725MG
Encompass_Preferred
Porcn-IN-1 is a potent small-molecule inhibitor of Porcupine (Porcn), an O-acyltransferase required for Wnt protein palmitoylation and secretion. It is supplied as a research reagent for in vitro and cell-based studies to probe Wnt signaling and porcupine-dependent biology; reported IC50 ≈ 0.5 ± 0.2 nM and purity ≈99.6%.
  • High potency against porcupine with low-nanomolar IC50.
  • High purity suitable for biochemical and cell-based assays.
  • Solid form with recommended solubility in DMSO for stock solutions.
  • Useful for probing Wnt signaling and related pathways in research models.
  • Available in small pack sizes for screening and follow-up experiments.
Catalog No. 50-002-63673