Learn More
ABclonal Technology PORCN Rabbit pAb
SDP
Supplier:  ABclonal Technology A122501000UL
Encompass_Preferred
This gene belongs to the evolutionarily conserved porcupine (Porc) gene family. Genes of the porcupine family encode endoplasmic reticulum proteins with multiple transmembrane domains. Porcupine proteins are involved in the processing of Wnt (wingless and int homologue) proteins. Disruption of this gene is associated with focal dermal hypoplasia, and the encoded protein has been implicated in cancer. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Catalog No. 50-001-14546