Learn More
ATCC Porphyromonas gingivalis (strain W50) - Anaerobic clinical bacterial culture
Supplier: ATCC BAA3229
Porphyromonas gingivalis (strain W50) is an anaerobic, gram-negative bacterial culture supplied as a freeze-dried clinical isolate for laboratory research and reference uses. Rehydrate the vial per instructions and culture under anaerobic conditions on enriched media at 37°C.
- Organism: Gram-negative anaerobe and clinical isolate.
- Strain designation: W50.
- Formulation: Freeze-dried culture in a vial.
- Rehydration: Rehydrate vial with 0.5 mL of broth before inoculation.
- Recommended media: Brucella blood agar or supplemented tryptic soy agar/broth with defibrinated sheep blood.
- Atmosphere: Anaerobic.
- Incubation temperature: 37°C.
- Storage: 2°C to 8°C.
- Biosafety: BSL 2 containment recommended.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.