Learn More
New Jersey Fluid Systems LLC PORT CONNECTOR
Supplier:  New Jersey Fluid Systems LLC SS401PC2
Encompass
316 Stainless Steel Swagelok Tube Fitting Reducing Port Connector. 1/4 in. OD x 1/8 in. OD
Catalog No. NC9954350