Learn More
Electron Microscopy Sciences Porta-Wand Kit for up to 6" (150mm) wafers, Hard anondized wafer tip and 115V In-stand Charger
SDP
Supplier:  Electron Microscopy Sciences 7192804
Encompass_Preferred
The Porta-Wand wafer handling toolkit comes with a non-removable 9.6V NiMH rechargeable battery and in-stand charger. Easy push-button on/off control. Replaceable internal air filter is accessible from the front of the tool. Internal exhaust filter ensures better than Class 1 performance. Indicator Light flashes when battery needs to be recharged, and also turn on when proper vacuum has been established. Comes with 115V in-stand charger.
Includes: Hard anondized wafer tip and 115V In-stand Charger
Catalog No. 50-193-1145