Learn More
Cayman Chemical Posaconazole 10mg
SDP
Supplier:  Cayman Chemical 1473710 MG
Encompass_Preferred
A broad-spectrum triazole antifungal agent active against various yeasts and molds, including Aspergillus, Candida, Fusarium, and Zygomycetes; inhibits the growth of fungi by inhibiting the lanosterol 14α-demethylase (IC50s = 7-200 nM), a key enzyme in the synthesis of ergosterol
Catalog No. 50-297-1620