Learn More
Medchemexpress LLC Posaconazole-d5 | 1217785-83-5 | MFCD11977890 | 99.1% | 705.81 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17373S5MG
Encompass_Preferred
Posaconazole-d5 is a deuterium-labeled form of Posaconazole. Posaconazole is a broad-spectrum, second-generation, triazole compound with antifungal activity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84228