Learn More
Medchemexpress LLC Poseltinib | 1353552-97-2 | 98.1% | 470.52 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10901010MG
Encompass_Preferred
Poseltinib is an orally active, selective, irreversible small molecule Bruton tyrosine kinase (BTK) inhibitor with an IC50 of 1.95 nM. It effectively inhibits signaling mediated by B cell receptors (BCR), Fc receptors (FcR), and Toll-like receptors (TLR). Poseltinib also exhibits anti-inflammatory activity and is being researched for rheumatoid arthritis.
  • Orally active, selective, irreversible small molecule Bruton tyrosine kinase (BTK) inhibitor.
  • Effectively inhibits signaling mediated by B cell receptors (BCR), Fc receptors (FcR), and Toll-like receptors (TLR).
  • Exhibits anti-inflammatory activity.
  • Potential for research in rheumatoid arthritis.
  • Appearance: solid, light yellow to yellow.
Catalog No. 50-003-46599