Learn More
Medchemexpress LLC HY-109010 5mg Medchemexpress, Poseltinib CAS:1353552-97-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090105MG
Encompass_Preferred
Medchemexpress, HY-109010 5mg Poseltinib CAS:1353552-97-2 Poseltinib, an orally active, selective and irreversible Bruton’s tyrosine kinase (BTK) inhibitor (IC50 =1.95 nM), with 0.3, 2.3 and 2.4-fold selectivity for BTK over BMX, TEC and TXK, respectively. Poseltinib can covalently bind to the active site (cysteine 481 residue) of BTK, and reveales potent inhibition of B cell receptor (BCR), Fc receptor (FcR), Toll-like receptor (TLR) mediated signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0758