Learn More
Medchemexpress LLC Posenacaftor (sodium) | 2095064-06-3 | 99.8% | 467.49 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY109187A10MM/1ML
Encompass_Preferred
Posenacaftor sodium is a cystic fibrosis transmembrane regulator (CFTR) protein modulator used in research for cystic fibrosis (CF). It acts as a CFTR corrector, designed to fix and restore the function of defective and misfolded CFTR protein, allowing it to move to the cell surface and function as a chloride channel, thereby helping to maintain fluid balance in the airways.
  • Corrects folding and trafficking of CFTR protein
  • Used for cystic fibrosis research
  • Restores function of defective CFTR protein
  • Helps maintain fluid balance in airways
Catalog No. 50-003-45372