Learn More
MSC PIP Cut, Puncture and Abrasion-Resistant Gloves, PolyKor/Aramid, 13 ga, 3X-Large, 12/Pk.
Supplier: MSC 00657387
These cut, puncture and abrasion-resistant gloves are designed for high-level protection in demanding industrial environments. Constructed with a 13 gauge seamless liner, they combine high-performance materials to provide reliable safety without compromising dexterity.
- ANSI/ISEA cut resistance level A6
- ANSI/ISEA abrasion resistance level 4
- ANSI/ISEA puncture resistance level 3
- Features a durable foam nitrile palm coating for enhanced grip
- Seamless lining composed of PolyKor and aramid fibers
- Knit cuff style ensures a secure fit
- EN 388 rating: 4X42FX