Learn More
American Radiolabeled Chemicals Inc POT BOROHYDRIDE 500LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1750500MC
Encompass_Preferred
Potassium borohydride-3H 500mCi as a solid, S.A. 70-100 Ci/mmol, M.W. 53.94, Exclusive
Catalog No. 50-901-186
Add to Cart
Request a Quote