Learn More
Cayman Chemical PotasIum PhosphAT Stock Sol 1L
SDP
Supplier:  Cayman Chemical 6002101 L
Encompass_Preferred
Potassium Phosphate Stock Solution (500 mM, pH 9.0) contains filtered sodium phosphate (500 mM). It is ready to use as supplied.
Catalog No. 50-289-4767