Learn More
Medchemexpress LLC Potassium 1H-indol-3-yl sulfate | 2642-37-7 | 99.9% | 251.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0119105MG
Encompass_Preferred
Potassium 1H-indol-3-yl sulfate is a tryptophan metabolite produced by intestinal microorganisms and sulfated in the liver before entering circulation. It acts as a potent endogenous agonist of the aryl hydrocarbon receptor (AhR) and is identified as a urinary toxin.
  • Used for research on kidney diseases
  • Significantly induces mRNA expression of CYP1A1/1A2/1B1 and UGT1A1/1A6
  • Regulates immune/inflammatory-related genes by continuously activating AHR
  • Exhibits plasma levels highly dependent on a healthy gut microbiota
Catalog No. 50-004-53822