Learn More
Strem, An Ascensus Company CAS# 13762-51-1. 50g. Potassium borohydride, 98%. MFCD00011396
SDP
Supplier:  Strem, An Ascensus Company 93190750G
Encompass
CAS# 13762-51-1. 50g. Potassium borohydride, 98%. MFCD00011396. Molecular Weight: 53.94. Molecular Formula: KBH4. Color/form: white pwdr. Strem# 93-1907. http://www.strem.com/catalog/v/93-1907/
Catalog No. 50-901-14258