Learn More
Medchemexpress LLC Potassium bromide | 7758-02-3 | MFCD00011358 | 500g
SDP
Supplier:  Medchemexpress LLC HYY1673500G
Encompass_Preferred
Potassium bromide 99% is a salt widely used as an anticonvulsant and a sedative Potassium bromide is a redox reagent that can be used to remove peripheral membrane proteins in molecular biology[1][2]
Catalog No. 50-002-66321