Learn More
Strem, An Ascensus Company CAS# 584-08-7. 500g. Potassium carbonate, 99+% (ACS). MFCD00011382
SDP
Supplier:  Strem, An Ascensus Company 931912500G
Encompass
CAS# 584-08-7. 500g. Potassium carbonate, 99+% (ACS). MFCD00011382. Molecular Weight: 138.21. Molecular Formula: K2CO3. Color/form: white pwdr. Strem# 93-1912. http://www.strem.com/catalog/v/93-1912/
Catalog No. 50-901-14264