Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium carbonate puriss1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 601091KGF
Encompass
Potassium carbonate is an inorganic base. It participates as a base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene.
Catalog No. 50-289-8549