Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium carbonate puriss500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 60109500GF
Encompass
Potassium carbonate is an inorganic base. It participates as a base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene.
Catalog No. 50-289-7496