Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium carbonate Reagen
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 31026312KG
Encompass
Potassium carbonate is an inorganic base. It participates as a base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene. Addition of amino acids (glycine sarcosine and proline) to K2CO3 (solvent) promotes the absorption of CO2.
Catalog No. 50-289-8917