Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium carbonate reagen25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3478252.5KG
Encompass
Potassium carbonate is an inorganic base. It participates as base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene.
Catalog No. 50-289-7614