Learn More
Quality Biological Inc Potassium Chloride, 1M, 500ML
SDP
Supplier:  Quality Biological Inc 351044101
Encompass_Preferred
Potassium Chloride 1 M is used to establish a constant ionic strength in a sample solution.KCl 74.56 g/LSuitable for PCR molecular biology protein chemistry and biochemical applications.Max Shelf Life: 36 months
Catalog No. 50-751-7650