Learn More
Boston Bioproducts Inc Potassium Chloride (2 M) - 100ml
Supplier:  Boston Bioproducts Inc MT252100ML
Encompass
The 2M Potassium Chloride Solution is an aqueous reagent that consists of potassium and chloride ions. When dissolved in water, these ions help the balance of electrolytes in living organisms and play a role in metabolic processes. When disscociated, KCl acts as a cofactor in enzymatic reactions related to protein synthesis and function. KCl Solution also enables biological assays, and is often used in the preparation of buffer solutions for polymerase chain reactions, DNA and RNA isolation.
Catalog No. NC1584098