Learn More
Apexbio Technology LLC Potassium Chloride 7447-40-7 1kg
SDP
Supplier:  Apexbio Technology LLC B72891000000
Encompass_Preferred
Potassium chloride (7447-40-7) is an inorganic salt that dissociates into potassium and chloride ions in aqueous solutions These ions function as electrolytes essential for maintaining osmotic balance regulating membrane potential and supporting signal transduction in cells Potassium chloride exerts its biological effects by supplying potassium ions thus restoring or maintaining physiological ionic conditions In experimental preparations it is widely utilized to formulate isotonic solutions or physiological buffers Potassium chloride is frequently applied in hypokalemia models cardiac electrophysiology studies and the preparation of nutrient media or buffering solutions for cell culture experiments
Catalog No. 50-000-91652