Learn More
Medchemexpress LLC Potassium chloride | 7447-40-7 | MFCD00011360 | 99.8% | 74.55 g/mol | KCl | 250g
SDP
Supplier:  Medchemexpress LLC HYY0537E250G
Encompass_Preferred
Potassium chloride for cell culture is potassium chloride that can be used for cell culture Potassium chloride for cell culture increases extracellular potassium ion concentration to induce cellular depolarization thereby activating voltage-gated calcium channels and regulating intracellular calcium signaling Potassium chloride for cell culture upregulates Brn-5 Potassium chloride for cell culture inhibits the proliferation of neurons and some plant cells[1][2]
Catalog No. 50-002-68994