Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium chloride ACS rea25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P39114X2.5KG
Encompass
Potassium chloride is an inorganic salt commonly used in fertilizer industry. Its generation via chlorination of microcline has been reported. The intrinsic and extrinsic conductivity of KCl crystals have been measured. The impact of KCl on the activity of mitochondrial membrane cristae and matrix proteins has been investigated. It facilitates the preparation of isoxazolines in aqueous medium.
Catalog No. 50-291-4636