Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium Chloride Primary
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NIST999C
Encompass
This Standard Reference Material (SRM) is intended for use as an analytical standard of known potassium (K) and chloride (Cl-) content. This lot of potassium chloride (KCl) was prepared to ensure a material of high purity and homogeneity and has been assayed after drying at 500 C. A unit of SRM 999c consists of a single glass bottle containing 30 g of the material.SRM 999C cert SRM 999C SDS
Catalog No. 50-290-6967