Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium chloride Reagent5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P45045KG
Encompass
Potassium chloride (KCl) is an inorganic salt commonly used in fertilizer industry. Its generation via chlorination of microcline has been reported. The intrinsic and extrinsic conductivity of KCl crystals have been measured. The impact of KCl on the activity of mitochondrial membrane cristae and matrix proteins has been investigated. It facilitates the preparation of isoxazolines in aqueous medium.
Catalog No. 50-290-2493