Learn More
Electron Microscopy Sciences Potassium Chloroplatinate Crystals
SDP
Supplier:  Electron Microscopy Sciences 80036
Encompass
Since crystal lattice plane spacings are accurately known from x-ray measurements, they provide a good test of microscope stability and a calibration of magnification in the upper range of magnification of the instrument. Plane Spacing 0.56
Catalog No. 50-268-84