Learn More
Medchemexpress LLC Potassium clavulanate cellulose | 98.0% | 238.26 (Potassium clavulanate) | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19964100MG
Encompass_Preferred
Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, functioning as a bacterial β-lactamase inhibitor. Clavulanate potassium, a form of clavulanic acid, is effective against bacteria resistant to penicillins and other antibiotics. When combined with amoxicillin, it can be utilized for research into various bacterial infections such as sinusitis, pneumonia, ear infections, bronchitis, urinary tract infections, and skin infections.
  • Functions as a bacterial β-lactamase inhibitor.
  • Effective against penicillin-resistant bacteria.
  • Can be combined with amoxicillin for bacterial infection research.
  • Inhibits various Bacteroides spp. and Fusobacterium spp.
  • Increases amoxicillin susceptibility rates for certain β-lactamase-positive bacterial species.
  • Shows protective effects in experimental pneumonitis models.
  • Appears as a white to off-white solid.
  • Recommended for research use only.
Catalog No. 50-004-03902