Learn More
Medchemexpress LLC Potassium clavulanate cellulose | 98.0% | 238.26 (Potassium clavulanate) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY199645MG
Encompass_Preferred
Potassium clavulanate cellulose is a mixture of potassium clavulanate and cellulose, functioning as a bacterial β-lactamase inhibitor. Clavulanate potassium, a form of Clavulanic acid, is effective against bacteria resistant to penicillins and other antibiotics. It can be utilized for research into various bacterial infections such as sinusitis, pneumonia, ear infections, bronchitis, urinary tract infections, and skin infections when combined with amoxicillin.
  • Acts as a bacterial β-lactamase inhibitor.
  • Combats bacteria resistant to penicillins and other antibiotics.
  • Used for research into bacterial infections including sinusitis, pneumonia, ear infections, bronchitis, urinary tract infections, and skin infections.
  • Increases amoxicillin susceptibility rates for β-lactamase-positive Bacteroides species and fusobacteria.
  • Protected mice with experimental pneumonitis in studies.
Catalog No. 50-004-05033