Learn More
Strem, An Ascensus Company CAS# 151-50-8. 250g. Potassium cyanide, 96% (ACS). MFCD00011397
SDP
Supplier:  Strem, An Ascensus Company 931916250G
Encompass
CAS# 151-50-8. 250g. Potassium cyanide, 96% (ACS). MFCD00011397. Molecular Weight: 65.12. Molecular Formula: KCN. Color/form: white pwdr. Strem# 93-1916. http://www.strem.com/catalog/v/93-1916/
Catalog No. 50-901-14268