Learn More
GFS Chemicals Inc POTASSIUM DICHROMATE, CERTIFIED | 500 G | GFS CHEMICALS, INC. Item #: 60602 | Bulk #: 78
SDP
Supplier:  GFS Chemicals Inc 60602
Encompass_Preferred
POTASSIUM DICHROMATE, CERTIFIED | Chemical Formula : K!2Cr!2O!7 | CAS # : 7778-50-9 | Specifications : Assay 99.95 - 100.05 %, Calcium 0.003 % , Chloride 0.001 % , Insoluble Matter 0.005 % , Iron 0.001 % , Loss on drying 0.05 % , NIST Traceable, Sodium 0.02 % , Sulfate 0.005 %
Catalog No. NC0099493