Learn More
Strem, An Ascensus Company CAS# 13967-50-5. 250mg. Potassium dicyanoaurate(I), 99%. MFCD00011414
SDP
Supplier:  Strem, An Ascensus Company 793000250MG
Encompass
CAS# 13967-50-5. 250mg. Potassium dicyanoaurate(I), 99%. MFCD00011414. Molecular Weight: 288.10. Molecular Formula: KAu(CN)2. Color/form: white pwdr. Strem# 79-3000. http://www.strem.com/catalog/v/79-3000/
Catalog No. 50-815-30