Learn More
Medchemexpress LLC Potassium dideuterium phosphate | 13761-79-0 | MFCD00074894 | 98.0% | 138.10 g·mol⁻1 | KD2PO4 | 500mg
SDP
Supplier:  Medchemexpress LLC HYY0332BS500MG
Encompass_Preferred
Phosphate monobasic-d2 (potassium) is the deuterium labeled Potassium phosphate monobasic[1] Potassium phosphate monobasic (DiA) is a commonly used in biological assay buffers Potassium phosphate monobasic is moderate to highly concentrated aqueous solutions of potassium phosphate monobasic for the production of phosphate buffers and other laboratory applications
Catalog No. 50-002-65895