Learn More
Strem, An Ascensus Company CAS# 7789-23-3. 500g. Potassium fluoride, anhydrous, 98%. MFCD00011398
SDP
Supplier:  Strem, An Ascensus Company 931922500G
Encompass
CAS# 7789-23-3. 500g. Potassium fluoride, anhydrous, 98%. MFCD00011398. Molecular Weight: 58.10. Molecular Formula: KF. Color/form: white pwdr. Strem# 93-1922. http://www.strem.com/catalog/v/93-1922/
Catalog No. 50-901-14278