Learn More
Strem, An Ascensus Company CAS# 13746-66-2. 250g. Potassium hexacyanoferrate(III), 99+% (ACS). MFCD00011392
SDP
Supplier:  Strem, An Ascensus Company 931920250G
Encompass
CAS# 13746-66-2. 250g. Potassium hexacyanoferrate(III), 99+% (ACS). MFCD00011392. Molecular Weight: 329.26. Molecular Formula: K3Fe(CN)6. Color/form: red xtl. Strem# 93-1920. http://www.strem.com/catalog/v/93-1920/
Catalog No. 50-901-14272