Learn More
Medchemexpress LLC Potassium hexacyanoferrate(III) | 13746-66-2 | MFCD00011392 | 99.5% | 329.24 g/mol | K3Fe(CN)6 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0998100G
Encompass_Preferred
Potassium hexacyanoferrate(III) 99 5% an iron-based cyanide complex is an oxidizing agent that can be used in the biocatalytic oxidation Potassium hexacyanoferrate(III) 99 5% can be used to prepare Glucose conductometric biosensor[1][2]
Catalog No. 50-002-68155