Learn More
Matrix Scientific Potassium hexafluorophosphate, 17084-13-8, MFCD00011412, 100g
Supplier: Matrix Scientific 001037100G
Molecular Formula F6KP, Purity 99%, Molecular Weight 184.07, Melting Point 575°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.