Learn More
Strem, An Ascensus Company CAS# 16871-90-2. 500g. Potassium hexafluorosilicate, 99%. MFCD00040666
SDP
Supplier:  Strem, An Ascensus Company 931959500G
Encompass
CAS# 16871-90-2. 500g. Potassium hexafluorosilicate, 99%. MFCD00040666. Molecular Weight: 220.28. Molecular Formula: K2SiF6. Color/form: white pwdr. Strem# 93-1959. http://www.strem.com/catalog/v/93-1959/
Catalog No. 50-901-14311