Learn More
Alkali Scientific Potassium Hydroxide, 5 Kg
SDP
Supplier:  Alkali Scientific SIU003CP
Encompass_Preferred
Potassium Hydroxide (5kg, Laboratory Grade) is a strong base used in a variety of industrial and laboratory applications. This bulk quantity is perfect for high-volume chemical processes, including neutralization reactions, saponification, and the preparation of potassium salts. Potassium hydroxide is frequently used in the manufacture of soaps, as it catalyzes the breakdown of fats and oils. Additionally, it is essential for preparing alkaline buffers and for pH adjustment in numerous chemical processes. Its high purity ensures consistency and reliability in laboratory testing and commercial production of potassium-based products.
Catalog No. 50-006-61594